Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

573822

Sigma-Aldrich

Anti-Synaptophysin Mouse mAb (SP15)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

biological source

mouse

Quality Level

form

liquid

species reactivity

(bovine, feline, hamster, human, mouse, porcine, rat)

storage condition

OK to freeze

isotype

IgM

storage temp.

−20°C

General description

Ammonium sulfate purified (50%) mouse monoclonal antibody. Recognizes the ~38 kDa syaptophysin protein.

Application

Frozen Sections (1:10; Immunoblotting (1:2000, colorimetric)Immunocytochemistry (1:250)Immunoprecipitation (1:100)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service