Skip to Content
MilliporeSigma

574796

TBE Buffer, 10X Powder Pack, ULTROL® Grade

Each pack provides an adequate amount of material to make 1 L of 10X concentrate that can be diluted to a 1X solution containing 89 mM Tris, 89 mM boric acid, 2 mM EDTA, pH ~8.3.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated

color

white

shipped in

ambient

storage temp.

10-30°C

General description

A commonly used electrophoretic buffer for the separation of DNA fragments. Each packet provides an adequate amount of material to make 1 liter of a 10X concentrate of buffer that can be diluted further. When diluted the 1X solution contains 89 mM Tris base, 89 mM boric acid, and 2 mM EDTA, pH 8.1-8.5.
Each pack provides an adequate amount of material to make 1 L of 10X concentrate that can be diluted to a 1X solution containing 89 mM Tris, 89 mM boric acid, 2 mM EDTA, pH ~8.3.

Preparation Note

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 2 months in the refrigerator (4°C). Discard the buffer solution if any growth appears.
Reconstitute each package in ~750-800 ml H₂O and mix until completely dissolved. Add enough H₂O to bring the final volume to 1 liter. The resulting solution is a 10X concentrate that can be further diluted to 1X as needed.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library