Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

575546

Sigma-Aldrich

Tankyrase1/2 Inhibitor III

The Tankyrase1/2 Inhibitor III controls the biological activity of Tankyrase1/2.

Synonym(s):

Tankyrase1/2 Inhibitor III, 3-(4-Methoxyphenyl)-5-((4-(4-methoxyphenyl)-5-methyl-4H-1,2,4-triazol-3-ylthio)methyl)-1,2,4-oxadiazole, TNKS1/2 Inhibitor III, Wnt Pathway Inhibitor XIV, TNKS1/2 Inhibitor III, Wnt Pathway Inhibitor XIV, 3-(4-Methoxyphenyl)-5-((4-(4-methoxyphenyl)-5-methyl-4H-1,2,4-triazol-3-ylthio)methyl)-1,2,4-oxadiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19N5O3S
CAS Number:
Molecular Weight:
409.46
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

CC1=NN=C(SCC2=NC(C3=CC=C(C=C3)OC)=NO2)N1C(C=C4)=CC=C4OC

InChI

1S/C20H19N5O3S/c1-13-22-23-20(25(13)15-6-10-17(27-3)11-7-15)29-12-18-21-19(24-28-18)14-4-8-16(26-2)9-5-14/h4-11H,12H2,1-3H3

InChI key

LQZSHPITKSPDLC-UHFFFAOYSA-N

General description

A cell-permeable triazolylthiomethyl-oxadiazole compound that binds TNKS1/PARP5a and TNKS2/PARP5b with high affinity (Kd = 79 and 28 nM, respectively), and acts as a potent and reversible TNKS1/2 dual inhibitor (IC50 = 33 nM TNKS2 in an auto-PARsylation assay). Shown to interact with the adenosine diphosphate linker portion of the NAD+ donor site, and display excellent selectivity over PARP1 and PARP2 activities (IC50 >19 µM); stabilize Axin2 levels (EC50 = 709 nM in SW480 cells), and antagonize Wnt signaling (IC50 = 215 nM in HEK293-SuperTopFlash assay). Exhibits minimal activity towards cytochrome P450 enzymes (IC50 = 21 and 5.7 µM for CYP2C9 and CYP3A4) and hERG ion channel (IC50 >30 µM), and adequate microsomal stability.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
TNKS1/PARP5a and TNKS2/PARP5b
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C. Avoid freeze/thaw cycles of solutions.

Other Notes

Shultz, M.D., et al. 2012. J. Med. Chem.55, 1127.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service