Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5910

Millipore

OmniPur® Lithium Chloride - CAS 7747-41-8 - Calbiochem

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

form

crystals

color

white

storage temp.

2-8°C

General description

Molecular biology grade.

Other Notes

Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service