MilliporeSigma
All Photos(1)

64969

Sigma-Aldrich

Krystalon Mounting Medium

HARLECO® Toulene-based. Use as a liquid coverslip or with a coverglass.

Synonym(s):
cover slipping

Quality Level

form

liquid

color

colorless

mp

-89.36 °C

transition temp

flash point 4.4 °C

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

10-30°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
O80151.172761.09016
Mounting Kit To upgrade of standard MAS-100 NT® and MAS-100 NT® Ex samplers with HEPA H13 filter

Millipore

1.17276

Mounting Kit

Neo-Mount™ anhydrous mounting medium for microscopy

Sigma-Aldrich

1.09016

Neo-Mount

color

colorless

color

clear

color

-

color

-

mp

-89.36 °C

mp

-

mp

-

mp

-

transition temp

flash point 4.4 °C

transition temp

-

transition temp

-

transition temp

flash point 19.5 °C

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

hematology
histology

application(s)

air monitoring
pharmaceutical

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

shipped in

-

shipped in

-

shipped in

-

Application

For In-Vitro Diagnostic Use.
For Professional Use.
HARLECO® Toulene-based. Use as a liquid coverslip or with a coverglass.

Packaging

64969-71: 500 mL glass bottle

64969-95: 6 x 50 mL glass bottles with application rod on bottle cap

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F

Flash Point(C)

4.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 5

1 of 5

Sigma-Aldrich

Sigma-Aldrich

317616

D.P.X.

DPX new non-aqueous mounting medium for microscopy

Sigma-Aldrich

1.00579

DPX new

Sigma-Aldrich

Sigma-Aldrich

C9368

CC/Mount

Goonho Park et al.
Cell reports, 31(13), 107839-107839 (2020-07-02)
Amyloid β-protein (Aβ) toxicity is hypothesized to play a seminal role in Alzheimer's disease (AD) pathogenesis. However, it remains unclear how Aβ causes synaptic dysfunction and synapse loss. We hypothesize that one mechanism of Aβ-induced synaptic injury is related to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service