MilliporeSigma
All Photos(1)

65068-M

Sigma-Aldrich

Gill Modified EA

HARLECO® EA stain modified to exclude Bismarck Brown

Synonym(s):
EA, Gill

Quality Level

form

liquid

mp

16.67 °C

transition temp

flash point 26 °C

density

0.79 g/cm3

λmax

528-538 nm (Eosin)
627-637 nm (Light Green)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
64817-U64815-U1.05175
Sigma-Aldrich

Sigma-Aldrich

65068-M

Gill Modified EA

Supelco

Supelco

64817-U

Condenser

Supelco

Supelco

64815-U

Condenser

mp

16.67 °C

mp

-

mp

-

mp

-

density

0.79 g/cm3

density

-

density

-

density

1.04-1.06 g/cm3 at 20 °C

λmax

528-538 nm (Eosin), 627-637 nm (Light Green)

λmax

-

λmax

-

λmax

-

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

food and beverages

application(s)

food and beverages

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

storage temp.

-

storage temp.

-

storage temp.

15-25°C

Application

HARLECO® EA stain modified to exclude Bismarck Brown
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Gill Modified EA (with Eosin and Light Green) modified to exclude Bismarck Brown.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Sens. 1 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

78.8 °F

Flash Point(C)

26 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service