65092D

Sigma-Aldrich

Iodine Solution Gram (Stabilized)

Harleco®, suitable for Gram staining

Synonym(s):
Gram Stain

storage conditions

Store at +15°C to +30°C.

Quality Level

form

liquid

manufacturer/tradename

Harleco®

application(s)

Gram staining: suitable
hematology: suitable
histology: suitable

mp

-0.1 °C

density

3.13 g/cm3

λmax

585-595 nm

Featured Industry

Diagnostic Assay Manufacturing

shipped in

ambient

storage temp.

15-25°C

General description

HARLECO® Individual replacement for the iodine component of the Gram Staining Set. Chemically binds crystal violet stain to the cell wall.

Packaging

4 l in Plastic bottle
500 ml in Plastic bottle

Application

HARLECO® Individual replacement for the iodine component of the Gram Staining Set. Chemically binds crystal violet stain to the cell wall.
Individual replacement for the iodine component of the Gram Staining Set. Chemically binds crystal violet stain to the cell wall.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Target Organs

Thyroid

WGK Germany

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.