65351-M

Sigma-Aldrich

Neo-Clear Xylene Substitute

Solvent used as a medium for clearing and deparafinizing steps in staining and tissue processing

Synonym(s):
Mineral Spirits
CAS Number:
EC Index Number:
232-489-3

storage conditions

Store at +15°C to +30°C.

Quality Level

application(s)

hematology: suitable
histology: suitable

color

clear

mp

-20 °C

transition temp

flash point 38 °C

Featured Industry

Diagnostic Assay Manufacturing

storage temp.

15-25°C

Application

Solvent used as a medium for clearing and deparafinizing steps in staining and tissue processing
Aromatic solvent used as a medium for clearing and deparafinizing steps in staining and tissue processing. Use with Neo-Mount coverslipping medium, Catalog #65038-71.

Legal Information

NEO-CLEAR is a trademark of Merck KGaA, Darmstadt, Germany
Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.