Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

676502

Sigma-Aldrich

InhibitorSelect VEGF Signaling Pathway Inhibitor Panel

The InhibitorSelect VEGF Signaling Pathway Inhibitor Panel controls the biological activity of InhibitorSelect.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A panel containing 15 potent and selective inhibitors useful for the study of VEGF signaling pathways. This panel contains the following inhibitors and 15 ml anhydrous DMSO for reconstitution:
A panel containing 15 potent, selective, reversible, and cell-permeable inhibitors useful for the study of multiple signaling pathways activated by VEGF: 1 mg of Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2 (Cat. No. 124018); 1 mg of Calcineurin Autoinhibitory Peptide, Cell-permeable (Cat. No. 207001); 5 mg of ET-18-OCH3 (Cat. No. 341207); 500 µg of Go 6983 (Cat. No. 365251); 100 mg of NG-Nitro-L-arginine Methyl Ester, Hydrochloride (Cat. No. 483125); 5 mg of p21-Activated Kinase Inhibitor III, IPA-3 (Cat. No. 506106); 500 µg of cPLA2α Inhibitor (Cat. No. 525143); 1 mg of PI-103 (Cat. No. 528100); 1 mg of PP2 (Cat. No. 529573); 1 mg of SB 203580 (Cat. No. 559389); 1 mg of U0126 (Cat. No. 662005); 5 mg of VEGFR Tyrosine Kinase Inhibitor V (Cat. No. 676501), 5 mg of VEGFR2 Kinase Inhibitor VI, Ki8751 (Cat. No. 676484); 1 mg of VEGFR2 Kinase Inhibitor III, SU5416 (Cat. No. 676487); 1 mg of ZM 336372 (Cat. No. 692000). Also provided with 15 ml of anhydrous DMSO (Cat. No. KP31817). Supplied with a data sheet.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) each inhibitor. Each inhibitor solution is stable for up to 3 months at -20°C, except 662005, which is unstable in solution, and should be reconstituted just prior to use.
Please refer to information provided on the CD for reconstitution information of individual inhibitors.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

188.6 °F - closed cup - (Dimethylsulfoxide)

flash_point_c

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service