Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

676503

Sigma-Aldrich

VEGFR/Tie-2/PDGFR Tyrosine Kinase Inhibitor

The VEGFR/Tie-2/PDGFR Tyrosine Kinase Inhibitor controls the biological activity of VEGFR/Tie-2/PDGFR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

VEGFR/Tie-2/PDGFR Tyrosine Kinase Inhibitor, 1-(2-Fluoro-4-((5-methyl-5H-pyrrolo[3,2-d]pyrimidin-4-yl)oxy)phenyl)-3-(3-(trifluoromethyl)phenyl)urea, PDGFR Tyrosine Kinase Inhibitor XXI, VEGFR Tyrosine Kinase Inhibitor XXXIV, VEGFR2 Kinase Inhibitor XXXII, VEGFR1 Kinase Inhibitor VII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H15F4N5O2
CAS Number:
Molecular Weight:
445.37
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

light beige

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(NC1=CC(C(F)(F)F)=CC=C1)NC2=CC=C(OC3=NC=NC4=C3N(C)C=C4)C=C2F

General description

An orally bioavailable pyrrolopyrimidinylurea compound that simultaneously binds to both the adenine and the adjacent hydrophobic pocket of the inactive form of VEGFR2 with high-affinity (type-II inhibitor) and acts as a pseudo-irreversible inhibitor with prolonged action. Shown to potently inhibit the activities of VEGFR2, VEGFR1, Tie-2, PDGFRα, PDGFRβ and c-kit (IC50 = 6.2, 15, 20, 35, 96 and 170 nM, respectively) with moderate to excellent selectivity over B-raf and Aurora-A (IC50 = 0.9 and 1.05 µM) and FGFR1, HER2, EGFR, IR, PKCθ (>10 µM) among 18-kinases. Arrests VEGF-induced HUVECs proliferation (IC50 = 4.4 µM) and suppresses DU145 carcinoma growth in a xenograft nude mouse model (6 mg/kg, b.i.d.).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 monthst at -20°C.

Other Notes

Oguro, Y., et al. 2010. Bioorg. Med. Chem.18, 7150.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service