Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

Skip To

681507

Anti-CaV1.2 (818-835) Rabbit pAb

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.41
UNSPSC Code:
51111800
Clone:
-
Species reactivity:
-
Application:
Citations:
-
This product is sold exclusively by Millipore via offline channels. Contact Customer Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

Storage Class

11 - Combustible Solids

wgk

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
681507-1ST04055977260700

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service