Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

682162

Sigma-Aldrich

Xestospongin D, Xestospongia sp.

A reversible, membrane-permeable blocker of IP3-mediated Ca2+ release (IC₅₀ = 648 nM).

Synonym(s):

Xestospongin D, Xestospongia sp., XeD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H50N2O3
Molecular Weight:
462.71
UNSPSC Code:
12352200

Quality Level

assay

≥90% (TLC)

form

film

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

clear

solubility

ethanol: 1 mg/mL
methanol: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A membrane-permeable, reversible blocker of IP3-mediated Ca2+ release (IC50 = 648 nM) that is less potent than XeC (Cat. No. 682160). Also displays a high selectivity over the skeletal isoform of the ryanodine receptor type 1 (Ry1R).
A reversible, membrane-permeable blocker of IP3-mediated Ca2+ release (IC50 = 648 nM). Also displays high selectivity over the skeletal isoform of the ryanodine receptor type 1 (RyR-1).

Biochem/physiol Actions

Cell permeable: no
Primary Target
IP3-mediated Ca2+
Product does not compete with ATP.
Reversible: yes
Target IC50: 648 nM in blocking IP3-mediated Ca2+ release

Warning

Toxicity: Harmful (C)

Preparation Note

Sonication may be required to achieve complete solubilization.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gafni, J., et al. 1997. Neuron 19, 723.
Tinker, A., and Williams, A.J. 1995. Biophys. J. 68, 111.
Callaway, C., et al. 1994. J. Biol. Chem. 269, 15876.
Quirion, J.C., et al. 1992. J. Nat. Prod. 55, 1505.
Nakagawa, M., et al. 1984. Tetrahedron Lett.25, 3227

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service