Skip to Content
MilliporeSigma

71229

pSTBlue-1 Perfectly Blunt Giga Cloning Kit

Sign In to View Organizational & Contract Pricing

Select a Size

This product is sold exclusively by Millipore via offline channels. Contact Customer Service

About This Item

UNSPSC Code:
41121800

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

shipped in

dry ice

Quality Level

storage temp.

−70°C

General description

Cloning kit with higher-efficiency competent cells The pSTBlue-1 Perfectly Blunt Giga Cloning Kits feature NovaBlue GigaSingles Competent Cells for increased transformation efficiency (> 1 × 109 cfu/µg). NovaBlue is a K-12 strain ideally suited as an initial cloning host due to its high transformation efficiency, blue-white screening capability (with appropriate plasmids) and recA endA mutations, which facilitate high yields of excellent-quality plasmid DNA. Cells are provided in a 50-µl, single-use volume that eliminates the need to dispense, freeze/thaw or waste partially used vials.

The Perfectly Blunt Cloning Kits are designed for simplified cloning of DNA generated by PCR using any type of DNA polymerase. This approach enables the use of high-fidelity proofreading enzymes for amplification, thus decreasing the probability of generating mutations in the target sequence. In addition, under many conditions blunt cloning is more efficient than T-cloning, most likely due to the observation that the efficiency of single dA addition by Taq DNA polymerase varies significantly depending on the sequence context of the DNA ends, and even the number of PCR cycles performed (1–5).

Note that the Perfectly Blunt method is not limited to cloning PCR products; these kits are also suitable for cloning restriction fragments, cDNA, or sheared DNA with the same protocols.

The pSTBlue-1 vector is available in a kit containing sufficient reagents for 20 or 40 reactions. "Vector only" kits are also available in 20 and 40 reaction sizes without ligase and competent cells.

Features and Benefits

  • No restriction enzymes or special primers
  • Compatible with any DNA polymerase
  • Independent of 3ʹ-dA addition or sequence context of DNA ends
  • Blue/white screening with seven different vectors, including pETBlue expression vectors
  • Simple protocol takes less than one hour from PCR product to plating transformants

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service