Skip to Content
MilliporeSigma

71339-M

pIEx-7 Ek/LIC Vector Kit - Novagen

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.54
UNSPSC Code:
41106617
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment



Pictograms

Exclamation mark

Storage Class

10 - Combustible liquids

WGK

WGK 3

Signal Word

Danger

Hazard Codes

Hazard Classifications

ED ENV 1 - Eye Irrit. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library