Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

7602

Sigma-Aldrich

Nuclear Fast Red (C.I. 60760)

HARLECO®General counterstain

Synonym(s):

Nuclear Fast Red, Helio Fast Rubin BBL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
357.27
Colour Index Number:
60760
MDL number:
UNSPSC Code:
41116130
EC Index Number:
229-088-0

Quality Level

form

solid

pH

5.5-7.0 (20 °C, 1 g/L in H2O)

solubility

1 g/L

λmax

542-552 nm

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Na+].[S](=O)(=O)(O)c1c(c2c(c(c1O)N)C(=O)c3c(cccc3)C2=O)[O-]

InChI

1S/C14H9NO7S.Na/c15-9-7-8(12(18)14(13(9)19)23(20,21)22)11(17)6-4-2-1-3-5(6)10(7)16;/h1-4,18-19H,15H2,(H,20,21,22);/q;+1/p-1

InChI key

IFSXZLJQEKGQAF-UHFFFAOYSA-M

Application

For In-Vitro Diagnostic Use.
For Professional Use.
General counterstain.
HARLECO®General counterstain

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service