Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

79198BC

Millipore

PPMX Surface Pro Power Cord EU

BioControl®, For use with GDS PickPen® PIPETMAX®

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$7.80

$7.80


Estimated to ship onJune 23, 2025Details


Request more information

Select a Size

Change View
1 EACH
$7.80

About This Item

UNSPSC Code:
41121521
NACRES:
NB.79

$7.80


Estimated to ship onJune 23, 2025Details


Request more information

Quality Level

storage condition

room temperature

technique(s)

PCR: suitable

application(s)

food and beverages

compatibility

for use with BioControl GDS

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
79199BC79113BC79114BC
technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

storage condition

room temperature

storage condition

room temperature

storage condition

-

storage condition

room temperature

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

-

Components

One EU Power Cord

Testing Method

PCR

Legal Information

BioControl is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service