8.45144

Sigma-Aldrich

Sodium butyrate

Msynthplus

Synonym(s):
Sodium butyrate, Butyric acid sodium salt
Empirical Formula (Hill Notation):
C4H7NaO2
CAS Number:
Molecular Weight:
110.09
MDL number:
EC Index Number:
205-857-6

storage conditions

Store below +30°C.

Quality Level

form

powder

mp

244 °C

density

1.324 g/cm3 at 30 °C

storage temp.

2-30°C

InChI

1S/C4H8O2.Na/c1-2-3-4(5)6;/h2-3H2,1H3,(H,5,6);/q;+1/p-1

InChI key

MFBOGIVSZKQAPD-UHFFFAOYSA-M

Analysis Note

Color (visual): white
Appearance of substance (visual): powder
Assay (HClO₄): ≥ 99.0 %
Identity (IR): passes test
1,1-Dichloroethene (HS-GC): lot specific result
1,1,1-Trichloroethane (HS-GC): lot specific result
Benzene (HS-GC): lot specific result
Carbon tetrachloride (HS-GC): lot specific result
1,2-Dichlorethane: not measurable

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

hazcat

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

storage_class_code

8A - Combustible, corrosive hazardous materials

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service