Skip to Content
MilliporeSigma

8.45147

1-Bromo-3-chloropropane

Synonym(s):

1-Bromo-3-chloropropane, 1-Chloro-3-bromopropane

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C3H6BrCl
CAS Number:
Molecular Weight:
157.44
UNSPSC Code:
12352101
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C3H6BrCl/c4-2-1-3-5/h1-3H2

InChI key

MFESCIUQSIBMSM-UHFFFAOYSA-N

vapor pressure

7.5 hPa ( 20 °C)

assay

≥99.0% (GC)

potency

1100-1200 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rat)

expl. lim.

3.2-8.6 % (v/v)

bp

141-143 °C/1013 hPa

mp

-59 °C

transition temp

flash point 76 °C

density

1.60 g/cm3 at 20 °C

storage temp.

2-30°C

Quality Level

Analysis Note

Appearance (visual): clear
Colour number: ≤ 50
Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.595 - 1.597
Water (K. F.): ≤ 0.10 %
Identity (IR): passes test

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Muta. 2 - Repr. 1A - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 2

flash_point_c

> 140 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service