MilliporeSigma
All Photos(1)

Documents

8.52232

Sigma-Aldrich

Fmoc-D-Phg-OH

Novabiochem®

Sign Into View Organizational & Contract Pricing

Synonym(s):
Fmoc-D-Phg-OH, N-α-Fmoc-D-phenylglycine
Empirical Formula (Hill Notation):
C23H19NO4
CAS Number:
Molecular Weight:
373.40
MDL number:

Quality Level

product line

Novabiochem®

assay

≥96.0% (acidimetric)
≥98% (TLC)
≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

170-180 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-30°C

InChI

1S/C23H19NO4/c25-22(26)21(15-8-2-1-3-9-15)24-23(27)28-14-20-18-12-6-4-10-16(18)17-11-5-7-13-19(17)20/h1-13,20-21H,14H2,(H,24,27)(H,25,26)/t21-/m1/s1

InChI key

PCJHOCNJLMFYCV-OAQYLSRUSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
8.522138.521488.53101-M
Fmoc-D-Phg-OH Novabiochem®

8.52232

Fmoc-D-Phg-OH

Fmoc-Phg-OH Novabiochem®

8.52213

Fmoc-Phg-OH

Fmoc-D-Phe-OH Novabiochem®

8.52148

Fmoc-D-Phe-OH

Boc-D-Phg-OH Novabiochem®

8.53101-M

Boc-D-Phg-OH

functional group

Fmoc

functional group

Fmoc

functional group

Fmoc

functional group

-

assay

≥96.0% (acidimetric), ≥98.0% (HPLC), ≥98% (TLC)

assay

≥96.0% (acidimetric), ≥98% (TLC), ≥98.0% (HPLC)

assay

≥97.0% (acidimetric), ≥98% (TLC), ≥99.0% (HPLC)

assay

≥98% (TLC)

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

-

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

mp

170-180 °C

mp

176-180 °C

mp

-

mp

88-91 °C

General description

Standard building block of introduction of D-phenylglycine amino-acid residues by Fmoc SPPS

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Linkage

Replaces: 04-13-1004

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 96.0 %
Water (K. F.): ≤ 1.0 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Articles

Unnatural amino acids, the non-proteinogenic amino acids that either occur naturally or are chemically synthesized, are becoming more and more important as tools for modern drug discovery research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service