MilliporeSigma
All Photos(1)

Documents

8.52288

Sigma-Aldrich

Fmoc-Phe(CF2PO3)-OH

Novabiochem®

Sign Into View Organizational & Contract Pricing

Synonym(s):
Fmoc-Phe(CF2PO3)-OH, Fmoc-F 2Pmp-OH,Fmoc-L-p-(phosphono-difluoromethyl)phenylalanine
Empirical Formula (Hill Notation):
C25H22F2NO7P
CAS Number:
Molecular Weight:
517.42
MDL number:

Quality Level

product line

Novabiochem®

assay

≥90.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

InChI

1S/C25H22F2NO7P/c26-25(27,36(32,33)34)16-11-9-15(10-12-16)13-22(23(29)30)28-24(31)35-14-21-19-7-3-1-5-17(19)18-6-2-4-8-20(18)21/h1-12,21-22H,13-14H2,(H,28,31)(H,29,30)(H2,32,33,34)/t22-/m0/s1

InChI key

XKWYQNFAGUNPJV-QFIPXVFZSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
8.522018.521958.52419
Fmoc-Phe(CF2PO3)-OH Novabiochem®

8.52288

Fmoc-Phe(CF2PO3)-OH

Fmoc-Phe-Thr(psiMe,Mepro)-OH Novabiochem®

8.52201

Fmoc-Phe-Thr(psiMe,Mepro)-OH

Fmoc-Phe-Ser(psiMe,Mepro)-OH Novabiochem®

8.52195

Fmoc-Phe-Ser(psiMe,Mepro)-OH

Fmoc-Cys(THP)-OH Novabiochem®

8.52419

Fmoc-Cys(THP)-OH

functional group

Fmoc

functional group

hydroxyl

functional group

hydroxyl

functional group

thiol

assay

≥90.0% (HPLC)

assay

≥97% (TLC), ≥97.0% (HPLC)

assay

≥97% (TLC), ≥97.0% (HPLC)

assay

-

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

15-25°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

General description

Building block [1] for the introduction by Fmoc SPPS of the hydrolytically stable phosphotyrosine analog phosphonodifluoromethylphenylalanine (F2Pmp) [2]. The pKa of F2Pmp more closely resembles that of a phosphate group making it better analog than phosphonomethylphenylalanine.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Guide to Selection of Orthogonally -Protected Building Blocks for Fmoc SPPS

Literature references

[1] M. F. Gordeev, et al. (1994) Tetrahedron Lett., 35, 7585.
[2] T. R. Burke, Jr. (2006) Curr. Top. Med. Chem., 6, 1465.

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Assay (HPLC, area%): ≥ 90.0 % (a/a)
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.1 %

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service