Skip to Content
MilliporeSigma

95-0004-06

BioTek® 405™ TSUVS Plate Washer for SMC® and Milliplex® Technology

Synonym(s):

Magnetic washer, Milliplex plate washer, SMC plate washer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.77
eCl@ss:
32160703
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


General description

SMC®/Milliplex® BioTek® 405 TS Washer with 96-well and 384-well magnetic wash capabilities

Application

SMC® bead-based assays, ELISA, magnetic bead based assays for Milliplex® for 96 and 384 wells

Features and Benefits

  • Self-maintaining design with an ultrasonic bath prevents clogging and failure in the plate washer. The built-in, patented Ultrasonic Advantage allows the instrument to clean build up in the manifold tube automatically, without manual intervention.
  • Hands-off, self-testing Verify technology eliminates tedious manual procedures using an ultrasonic probe to check dispense and aspirate performance. Test results are automatically displayed and inidicate when tubes may require additional cleaning.
  • Automated magnetic and filtration bead-based assays provide flexible operation in many applications. Variable wash steps and magnet accessories for biomagnetic separation workflows ensure high bead recovery. Vacuum filtration module is also available.
  • Cell-friendly design for high-quality results with angled dispense tubes, highly adjustable dispense and aspiratoin rate settings for cell-based asssays. The gentle flow rates and side-wall dispensing position ensure the integrity of cell monolayers.
  • SMC® custom magnet serialized to plate washer for optimal plate washing and performance for SMC® assays, including customized wash protocols.
  • Adaptible for MILLIPLEX® with Dexter magnets (purchased separately) and customized wash protocols.

Other Notes

Using 96 and 384-well Dexter magnets (sold separately), the washer is easily adapable for use with MILLIPLEX ®magnetic bead assays with the incorporation of customized MILLIPLEX® wash protocols pre-programmed on the washer.
When counting at the single molecule level, it is critical to ensure reliable assay performance. For SMCS® assays, one vital consideration is to ensure thorough bead washing during key stages of the workflow. The BioTek® 405 TSUVS Plate Washer for SMC® technology has been specifically modified with a custom magnetic plate and SMC® specific wash procedures to improve assay performance by preventing bead loss and ensuring consistency between wells.

Legal Information

BioTek is a registered trademark of BioTek Instruments LLC
405 is a trademark of Agilent Technologies, Inc.
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
SMC is a registered trademark of Singulex




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library