Skip to Content
MilliporeSigma

A007

Millipore

NovaSeptum Bag rack, multi sampling, for 1-port Holder TC25, in-line

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
41104105
eCl@ss:
36101790

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

sterility

sterile

Quality Level

H

260-460 mm

L

250 mm

needle size

2.0 mm

width

280 mm

weight

0.5 kg1.1 lb

suitability

suitable for sampling


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service