Skip to Content
MilliporeSigma

ABF1059

Sigma-Aldrich

Anti-MxB Antibody

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

This product is sold exclusively by Millipore via offline channels. Contact Customer Service

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

conjugate

unconjugated

Quality Level

antibody form

serum

clone

polyclonal

purified by

(unpurified)

species reactivity

human, rabbit

concentration

(Please refer to lot specific datasheet.)

technique(s)

western blot: suitable

UniProt accession no.

target post-translational modification

unmodified

General description

GST-tagged recombinant protein corresponding to the Dynamin-type G domain of human MxB.
~78 kDa observed

Application

Epitope: Dynamin-type G domain

Biochem/physiol Actions

Expected to react with both spliced isoforms.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.

Preparation Note

Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in IFN alpha 2b treated SupT1 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antiserum detected IFNalpha-2b-induced MxB/Mx2 expression in human T lymphoblast SupT1 cells.

Other Notes

Human. Predicted to react with Primate based on 100% sequence homology.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service