Skip to Content
MilliporeSigma

ABS2238

Sigma-Aldrich

Anti-SLC25A44

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352203

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

clone

polyclonal

Quality Level

isotype

IgG

Immunogen

KLH-conjugated linear peptides corresponding to 20 amino acids from the N-terminal region, 14 amino acids from the internal region, and 14 amino acids from the C-terminal region of mouse Solute carrier family 25 member 44 (SLC25A44).

Biochem/physiol Actions

This rabbit polyclonal antibody detects Solute carrier family 25 member 44 (SLC25A44) in multiple species.

Preparation Note

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Stable for 1 year at +2°C to +8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service