Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABT545

Sigma-Aldrich

Anti-Aggrecan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

antibody form

purified antibody

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rabbit, mouse, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

General description

GST/His-tagged recombinant fragment corresponding to 150 amino acids from the C-terminal half of murine Aggrecan core protein.

Specificity

Anti-Aggrecan is a rabbit polyclonal antibody that detects Aggrecan core protein. It targets an epitope within 150 amino acids from the C-terminal half.

Application

Epitope: unknown

Quality

Evaluated by Western Blotting in human Chondrocytes.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Aggrecan in lysate from human chondrocytes.

Physical properties

~270 kDa observed; 221.94 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Human, Mouse

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service