Skip to Content
MilliporeSigma

ACS396

Hydrofluoric Acid

Meets ACS Specifications

Synonym(s):

Hydrogen Fluoride, Hydrofluoric acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352301
Concentration:
48.0-51.0% Hf
Form:
liquid
Grade:
ACS reagent
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

form

liquid

shelf life

24 mo. (from mfg date)

concentration

48.0-51.0% Hf

impurities

≤0.01% Fluosilicic acid (H2SiF4), ≤0.05 ppm Arsenic (As), ≤0.1 ppm Copper (Cu), ≤0.5 ppm Heavy metals (as Pb), ≤1 ppm Iron (Fe), ≤1 ppm Phosphate (PO4), ≤5 ppm Chloride (Cl), ≤5 ppm Sulfate and sulfite (as SO4)

ign. residue

≤5 ppm

bp

106 °C/1013 hPa

mp

-35 °C

shipped in

ambient

storage temp.

no temp limit

Application

Meets ACS Specifications

Analysis Note

Arsenic (As): 0.05ppm max
Assay: 48.0-51.0%
Chloride (Cl): 5ppm max
Copper (Cu): 0.1ppm max
Expiration date: Exp 24 months from receipt date
Fluosilicic acid (H2SiF4): 0.01% max
Heavy metals (as Pb): 0.5ppm max
Iron (Fe): 1ppm max
Phosphate (PO4): 1ppm max
Residue after ignition: 5ppm max
Sulfate and sulfite (as SO4): 5ppm max


Still not finding the right product?

Explore all of our products under Hydrofluoric Acid


pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library