Skip to Content
MilliporeSigma

AP124X4

IGG H&L, Gt x Ms, AP >/= 4 mg/ml

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

eCl@ss:
32160702
UNSPSC Code:
12352203
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

antibody form

affinity purified VHH antibody

clone

monoclonal

species reactivity

mouse

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

Quality Level

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service