Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
NACRES:
NA.41
UNSPSC Code:
51111800
Species reactivity:
human
Species reactivity:
-
Application:
—
Citations:
-
biological source
mouse
form
liquid
species reactivity
human
storage condition
OK to freeze
dilution
(Frozen Sections
Immunoblotting
Immunoprecipitation
Paraffin Sections )
isotype
IgG
storage temp.
−20°C
Quality Level
General description
Contains 20 µg of each antibody. Includes Anti-MMP-1 (Cat. No. IM35), Anti-MMP-2 (Cat. No. IM33), Anti-MMP-3 (Cat. No. IM36), Anti-MMP-9 (Cat. Nos. IM09 and IM37), Anti-TIMP-1 (Cat. No. IM32), and Anti-TIMP-2 (Cat. No. IM11). Please see individual product descriptions for more information. The antibodies to MMP-1, MMP-2, and MMP-3 recognize both latent and active forms.
Application
Frozen Sections Immunoblotting Immunoprecipitation Paraffin Sections
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service