Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AX1699B

Supelco

Solvent S

AQUASTAR®

Synonym(s):

Karl Fischer

Sign Into View Organizational & Contract Pricing


About This Item

Quality Level

form

liquid

transition temp

flash point 15.6 °C

shipped in

ambient

storage temp.

room temp

Application

AQUASTAR
AX1699B-6, AX1699B-3 and AX1699B-5 are packaged in 38/430 poly coated glass bottles

Analysis Note

Color: Colorless to yellow
Form: Clear liquid
Performance: To pass test
Water (H2O): 0.001-0.01%

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT RE 2 Oral - STOT SE 1

target_organs

Eyes,Central nervous system, Kidney,Liver,Blood,Thyroid, Lungs

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

60.1 °F

flash_point_c

15.6 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service