Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BX1627

Supelco

Buffer Concentrate, pH 7.0

Synonym(s):

buffers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161703

grade

analytical standard

Quality Level

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

shipped in

ambient

storage temp.

15-25°C

General description

Buffer concentrate, pH 7.0 is an off-the-shelf buffer solution used in standardized method applications such as pH calibration.

Application

This ready-to-use pH 7.0 buffer solution comprising the phosphate mixture is suitable for use in pH meter calibration.

Features and Benefits

  • Custom-tailored solution for a cost-effective process with minimized risk
  • Stored in a plastic bottle equipped with a convenient squirt top dispensing cap
  • Traceable to NIST standards
  • Safety labeled
  • Recommended for use in general chemistry applications

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service