Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DX2113

Supelco

1,3-Dioxolane

Synonym(s):

Dioxolane

Sign Into View Organizational & Contract Pricing


About This Item

vapor pressure

133 hPa ( 20 °C)

Quality Level

assay

≥99% (GC)

form

liquid

impurities

≤0.001% Peroxide (as H2O2)
≤0.05% Water (H2O)

evapn. residue

≤0.001%

color

APHA: ≤20

bp

74-75 °C/1013 hPa

mp

-95 °C

transition temp

flash point -6 °C

solubility

1000 g/L

storage temp.

room temp

Analysis Note

Assay (GC):99% min
Color (APHA):20 max
Evaporation residue:0.001% max
Form:Clear liquid
Identity (IR-spectrum):Conforms
Peroxide (as H2O2):0.001% max
Water (H2O):0.05% max

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

26.6 °F - closed cup

flash_point_c

-3 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service