Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DX2415

Supelco

Dodecane

Synonym(s):

Dodecane, extraction

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CH3
CAS Number:
MDL number:
UNSPSC Code:
12352001
EC Index Number:
203-967-9
assay:
≥99% (GC)
bp:
216.3 °C/1013 hPa
vapor pressure:
0.4 hPa ( 20 °C)

vapor pressure

0.4 hPa ( 20 °C)

Quality Level

assay

≥99% (GC)

form

liquid

color

APHA: ≤20

bp

216.3 °C/1013 hPa

mp

-9.6 °C

transition temp

flash point 85.5 °C

density

0.75 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

InChI

1S/C12H26/c1-3-5-7-9-11-12-10-8-6-4-2/h3-12H2,1-2H3

InChI key

SNRUBQQJIBEYMU-UHFFFAOYSA-N

Analysis Note

Assay (GC): 99% min
Color (APHA): 20 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

supp_hazards

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

158.0 °F - Pensky-Martens closed cup

flash_point_c

70 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service