E711-10050
NovaSeptum® GO Sampling Unit; High Purity Application

Single; 50mL
Select a Size
$4,590.00
Select a Size
About This Item
Skip To
$4,590.00
material
316 stainless steel cannula (ASTM®)
PureFlex™ bag
TPE tubing (Thermoplastic elastomer)
polyester body
silicone septum (platinum-cured)
Quality Level
agency
ISO 10993-5 (all component materials)
USP 87 Cytotoxicity Elution Test (In Vitro)
according to ISO 11137 (sterilization)
according to ISO 146441
meets the criteria for Biological Reactivity Testing: can be any one or a combination of the following test methods USP<88> Systemic Injection Test (In Vivo)
sterility
sterile; β-irradiated
product line
NovaSeptum® GO
feature
autoclavable: no
parameter
-20-50 °C temp. range (-4-122 °F) (up to -80°C/-112°F when used with a NovaSeptum® Case)
0.50 bar max. pressure (7.25 psi) (up to 250 mL)
impurities
<2.15 EU/device bacterial endotoxins (LAL test)
fitting
female Luer-Lok® outlet (3-piece)
male Luer-Lok® outlet (3-piece containing an injection site)
application(s)
cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pathogen testing
pharma/biopharma processes
pyrogen testing
sterile sampling
sterility testing
viral therapy
storage temp.
room temp
General description
Application
Sterility testing, pyrogenic testing, endotoxin testing, chemical analysis, pH analysis, and fermentation applications.
Packaging
Components
- Septum: Platinum-cured silicone
- Body: Polyester
- Cannula: ASTM® 316 L Stainless steel
- Fluid contact layer (bag): Polyethylene film (PureFlex™ film)
- Tubing: Thermoplastic elastomer (TPE)
- Outlet Tubing: 3-piece Luer-Lok®, containing a male, female and an injection site
Preparation Note
Aqueous extraction contains < 2.15 EU per device as determined using the Limulus Amebocyte Lysate (LAL) test.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service