Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E714-10100

NovaSeptum® GO Sampling Unit; High Purity Application

new

Manifold; 5x100mL

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$2,826.00

$2,826.00


Check Cart for Availability


Select a Size

Change View
100 ML
$2,826.00

About This Item

UNSPSC Code:
41104105

$2,826.00


Check Cart for Availability

material

316 stainless steel cannula (ASTM®)
PureFlex polyethylene bag
TPE tubing (Thermoplastic elastomer)
polyester body
silicone septum (platinum-cured)

Quality Level

agency

ISO 10993-5 (all component materials)
USP 87 Cytotoxicity Elution Test (In Vitro)
according to ISO 11137 (sterilization)
according to ISO 146441
meets the criteria for Biological Reactivity Testing: can be any one or a combination of the following test methods USP<88> Systemic Injection Test (In Vivo)

sterility

sterile; β-irradiated

product line

NovaSeptum® GO

feature

autoclavable: no

parameter

-20-50 °C temp. range (-4-122 °F) (up to -80°C/-112°F when used with a NovaSeptum® Case)
0.30 bar max. pressure (4.35 psi)

impurities

<2.15 EU/device bacterial endotoxins (LAL test)

fitting

female Luer-Lok® outlet (3-piece)
male Luer-Lok® outlet (3-piece containing an injection site)
male Luer-Lok® outlet (3-piece)

application(s)

cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pathogen testing
pharma/biopharma processes
pyrogen testing
sterile sampling
sterility testing
viral therapy

storage temp.

room temp

General description

A closed, sterile sampling method can significantly reduce risk of cross contamination. Ideal for sampling from aseptic and sterile processes, the NovaSeptum® GO sterile sampling system provides consistently representative samples, safely and securely. NovaSeptum® GO high purity bags for multi-sampling systems are available from 50 to 250 mL.
Device configuration: Multi-sampling

Application

Regular Sampling Sample (with or without protective case).
Sterility testing, pyrogenic testing, endotoxin testing, chemical analysis, pH analysis, and fermentation applications.

Packaging

5x100 ml bags, 5 units per box

Components

  • Septum: Platinum-cured silicone
  • Body: Polyester
  • Cannula: ASTM® 316 L Stainless steel
  • Fluid contact layer (bag): Polyethylene film (PureFlex film)
  • Tubing: Thermoplastic elastomer (TPE)
  • Outlet Tubing: 3-piece Luer-Lok®, containing a male, female and an injection site

Preparation Note

Units are integrity tested at regular intervals during manufacturing.
Aqueous extraction contains < 2.15 EU per device as determined using the Limulus Amebocyte Lysate (LAL) test.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
Luer-Lok is a registered trademark of Becton-Dickinson & Co.
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany
PUREFLEX is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service