E871-80060
NovaSeptum® GO Sampling Unit; Bottle Application

Single; 60mL
Select a Size
$5,322.00
Select a Size
About This Item
Skip To
$5,322.00
material
316 stainless steel cannula (ASTM®)
PETG bottle
PureFlex™ polyethylene bottle
TPE tubing (Thermoplastic elastomer)
polyester body
silicone septum (platinum-cured)
Quality Level
agency
ISO 10993-5 (all component materials)
USP 87 Cytotoxicity Elution Test (In Vitro)
according to ISO 11137 (sterilization)
according to ISO 146441
meets the criteria for Biological Reactivity Testing: can be any one or a combination of the following test methods USP<88> Systemic Injection Test (In Vivo)
sterility
sterile; γ-irradiated
product line
NovaSeptum® GO
feature
autoclavable: no
parameter
-80-50 °C temp. range (-112-122 °F)
0.50 bar max. pressure (7.25 psi)
impurities
<2.15 EU/device bacterial endotoxins (LAL test)
fitting
outlet (Cap)
application(s)
bioburden testing
cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pathogen testing
pharma/biopharma processes
sterile sampling
viral therapy
storage temp.
room temp
General description
Application
Bioburden-Archiving
Packaging
Components
- Septum: Platinum-cured silicone
- Body: Polyester
- Cannula: ASTM® 316 L Stainless steel
- Fluid contact layer (bottle): Polyethylene Terephtalate Glycol(PETG)
- Tubing: Thermoplastic elastomer (TPE)
- Outlet Tubing: Cap
Preparation Note
Aqueous extraction contains < 2.15 EU per device as determined using the Limulus Amebocyte Lysate (LAL) test.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service