Skip to Content
MilliporeSigma

E874-80250

NovaSeptum® GO Sampling Unit; Bottle Application

new

Manifold; 5x250mL

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

5 X 250 ML
$2,062.00

$2,062.00


Check Cart for Availability

Select a Size

Change View
5 X 250 ML
$2,062.00

About This Item

UNSPSC Code:
41104105

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

$2,062.00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

316 stainless steel cannula
PureFlex polyethylene bag (drainage bag)
TPE tube
polyester body
polyethylene bottle (fluid contact layer)
silicone septum (platinum-cured)

Quality Level

agency

ISO 10993-5 (all component materials)
USP 87 Cytotoxicity Elution Test (In Vitro)
according to ISO 11137 (sterilization)
according to ISO 146441
meets the criteria for Biological Reactivity Testing: can be any one or a combination of the following test methods USP<88> Systemic Injection Test (In Vivo)

sterility

sterile; γ-irradiated

product line

NovaSeptum® GO

feature

autoclavable: no

parameter

-80-50 °C temp. range (-112-122 °F) (Freeze sampling bottle vertically)
0.30 bar max. pressure (4.35 psi)

impurities

<2.15 EU/device bacterial endotoxins (LAL test)

application(s)

bioburden testing
cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pathogen testing
pharma/biopharma processes
sterile sampling
viral therapy

storage temp.

room temp

General description

Device configuration: Multi-sampling
The NovaSeptum® GO bottle sampling unit is intended for applications where high purity, such as low endotoxin levels, is required. This sampling unit is available with a 2 mm needle and must be used with a NovaSeptum® GO holder. The bio-neutral plastic bottles are ideal for all tests and available in a range of sizes. A closed, sterile sampling method can significantly reduce risk of cross contamination. Ideal for sampling from aseptic and sterile processes, the NovaSeptum® GO sterile sampling system provides consistently representative samples, safely and securely.

Application

Endotoxin
Bioburden-Archiving

Packaging

5x250 ml bottles, 3 units per box

Components

  • Septum: Platinum-cured silicone
  • Body: Polyester
  • Cannula: ASTM® 316 L Stainless steel
  • Fluid contact layer (bottle): Polyethylene Terephtalate Glycol
  • (Drainage bag): PureFlex Polyethylene filmfilm)
  • Tubing: Thermoplastic elastomer (TPE)
  • Outlet Tubing: Cap

Preparation Note

Units are integrity tested at regular intervals during manufacturing.
Aqueous extraction contains < 2.15 EU per device as determined using the Limulus Amebocyte Lysate (LAL) test.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany
PUREFLEX is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service