MilliporeSigma
EZFKIT001WW-M
Millipore
EZ-Fluo System standard kit including 1 reader, 1 camera and installation CD
Slide 1 of 1
About This Item
UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NB.79
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
product line
EZ-Fluo
Quality Level
General description
EZ-Fluo is a non-destructive method that enables you to continue to grow the microorganisms after they have been stained in order to identify them using any standard ID technology.
Application
EZ-Fluo is a non-destructive method that enables you to continue to grow the microorganisms after they have been stained.
Legal Information
EZ-FLUO is a trademark of Merck KGaA, Darmstadt, Germany