Key Documents
EZFREAD01WW
Millipore
EZ-Fluo reader
About This Item
UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NA.85
Pricing and availability is not currently available.
product line
EZ-Fluo
Quality Level
L
24.9 cm
weight
4.4 kg9.7 lbs
General description
EZ-Fluo is a non-destructive method that enables you to continue to grow the microorganisms after they have been stained in order to identify them using any standard ID technology.
Application
EZ-Fluo is a non-destructive method that enables you to continue to grow the microorganisms after they have been stained.
Legal Information
EZ-FLUO is a trademark of Merck KGaA, Darmstadt, Germany