GC0062

Supelco

Hydrochloric Acid .5N

.5N

Synonym(s):
Hydrogen chloride solution, Hydrochloric acid solution, HCl
Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:

storage conditions

No declaration

Quality Level

pH

<1 (20 °C in H2O)

shipped in

ambient

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Packaging

20 l in Plastic container

Application

.5N

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

WGK Germany

WGK 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.