Skip to Content
MilliporeSigma

GF444

Sigma-Aldrich

Vascular Endothelial Growth Factor 121 human

HumanKine®, recombinant, Xeno-free, >95% (SDS-PAGE), suitable for cell culture

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

This product is sold exclusively by Millipore via offline channels. Contact Customer Service

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

HumanKine® VEGF121, Human Recombinant Xeno-Free,

assay

>95% (SDS-PAGE)

Quality Level

feature

endotoxin free

UniProt accession no.

General description

HumanKine growth factors are expressed in a proprietary human 293 protein expression system and are manufactured without the use of any animal products and are considered Xeno-Free.
Vascular endothelial growth factor (VEGF) is active in angiogenesis, vasculogenesis and endothelial cell growth. VEGF induces endothelial cell proliferation, promotes cell migration, inhibits apoptosis, and induces permeabilization of blood vessels. VEGF binds to the VEGFR1/Flt-1 and VEGFR2/Kdr receptors, heparan sulfate and heparin. VEGF121 is acidic and is a freely secreted form of VEGF.

Physical form

Product is presented in PBS and filtered through a 0.2 micron filter before lyophilization.

Preparation Note

Store at -20°C for up to 4 months from date of receipt. Further dilutions should be made into appropriate buffer or medium to which protein has been added.

Analysis Note

Bioactivity assay: Determined by the dose-dependent stimulation of the proliferationof HUVEC cells (Human Umbilical Vein
Endothelial Cells). ED50 is typically 2-8 ng/ml.
HumanKine VEGF121 is manufactured using all non-animal reagents and is considered Xeno-Free

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service