All Photos(1)

GFCP0010

Millipore

G041 Glass Fiber Conjugate Pad Rolls

Lateral Flow, Assay Development, lateral flow immunoassays, lateral flow assay device, Immunoassay IVDs

eCl@ss:
32031602
NACRES:
NB.23

Quality Level

manufacturer/tradename

SureWick®

L

100 m (328 ft)

W

10 mm

filter L × W

1.0 cm × 100 m

slit W

10 cm

Related Categories

Application

Lateral Flow, Assay Development, lateral flow immunoassays, lateral flow assay device, Immunoassay IVDs

Legal Information

SUREWICK is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service