The product appears to consist of many strands of fiber rather than a sheet. It resembles an item shown on the website provided.
GX0090
Glass Wool
Synonym(s):
Glass Wool, Fibrous Glass
impurities
≤0.005% Heavy metals (as Pb)
≤0.1% Alkali soluble in water (as Na2O)
≤1% Substances soluble in hydrochloric acid
pH
9-11 (20 °C, 100 g/L in H2O, slurry)
shipped in
ambient
storage temp.
room temp
InChI
1S/Ca.Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q+2;+1;/p-3
InChI key
QXJJQWWVWRCVQT-UHFFFAOYSA-K
Application
- Assessment of different factors on the influence of glass wool concentration for detection of main swine viruses in water samples: This research demonstrates the use of glass wool in the filtration and concentration of viruses from environmental samples, contributing significantly to public health safety and environmental monitoring (Fan et al., 2023).
- The Evolution of Insulation Performance of Fiber-Reinforced Silica Aerogel after High-Temperature Treatment: Research on glass wool′s effectiveness in thermal insulation and its durability under high temperatures is essential for its application in high-performance building envelopes and industrial installations (Gao et al., 2023).
Analysis Note
signalword
Danger
hcodes
Hazard Classifications
Carc. 1B Inhalation
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Can you please tell me if the appearance of this glass wool (GX0090-2) is a sheet?
1 answer-
Helpful?
-
-
Can you tell me the shelf life of the glass wool? We have a box of the GX0090-2 that we purchased in 2018 and I am wondering when it would need to be replaced.
1 answer-
This material will have a lot specific expiration date which will be provided on the certificate of analysis. The expiration date for this material has about a 5 year window. Please visit this product page to download the certificate of analysis: https://www.emdmillipore.com/product/Glass-Wool,EMD_CHEM-GX0090#anchor_COA
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service