GX0205

Supelco

Glycine

GR ACS

Synonym(s):
Aminoacetic acid, Aminoacetic Acid, Aminoethanoic acid, Glycocoll
Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein/REAXYS Number:
635782
MDL number:
EC Index Number:
200-272-2

storage conditions

Store at room temperature.

Quality Level

vapor pressure

0.0000171 Pa ( 25 °C)

assay

≥98.5%

pH

5.9-6.4 (20 °C, 50 g/L in H2O)

pKa (25 °C)

(1) 2.35, (2) 9.60
2.35

mp

240 °C (dec.) (lit.)

solubility

soluble 250 g/L

shipped in

ambient

storage temp.

room temp

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

GR ACS Glycine
GX0205-1 1kg Glass Bottle

Packaging

1 kg in Plastic bottle

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Analysis Note

Ammonium (NH4): 0.005% max
Assay: 98.5% min
Chloride (Cl): 0.005% max
Heavy metals (as Pb): 0.002% max
Hydrolyzable substances: Passes test
Residue after ignition: 0.1% max
Substances darkened by sulfuric acid: Passes test
Sulfate (SO4): 0.005% max

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.