Skip to Content
MilliporeSigma

HCS30

BrdU Immunohistochemistry System

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.23
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


signalword

Danger

target_organs

Central nervous system, Kidney, Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 3

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Muta. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation - STOT RE 2 Oral - STOT SE 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library