Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HTS190RTA

Sigma-Aldrich

Ready-to-Assay NK3 Tachykinin Receptor Frozen Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

description

Cell line type: GPCR Cell Lines
GPCR Class A
protein target NK3

Quality Level

technique(s)

calcium flux assay: suitable

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

NK3 is one of three mammalian G-protein coupled receptors subtypes (NK1, NK2, NK3) that mediate the biological effects of the tachykinin peptide family. Neurokinin B is the tachykinin family peptide with the highest affinity to NK3R, but it still induces a response with NK1, and NK2 with lower potency (Pennefather et al., 2004; Regoli et al., 1994; Maggi et al., 1993). NK3, like the other tachykinin receptors, signals through Gq to stimulate intracellular calcium (Pinnock et al., 1994). Mammalian tachykinins are present in the brain and gut. NK3 receptor antagonists reduce intestinal motility and nociception in animal models of gastrointestinal disorders, and may be effective in irritable bowel syndrome (Sanger 2004). Millipore′s cloned human NK3-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant NK3 expression on the cell surface and contains high levels of the promiscuous G protein Gα15 to enhance coupling the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for antagonists of interactions between NK3 and its ligands.

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial. Fifty (50) mL of Media Component.

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service