Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LD0HCH6S6

Millipore

Millistak+® Lenticular Disc Filters, D0HC media series, 16 in. diameter, 2-cell, silicone seal, 6/pack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690
Pricing and availability is not currently available.

material

cellulose
mixed cellulose esters (HA)
polypropylene
silicone seal

Quality Level

description

sanitization May be hot water sanitized for 20 cycles using purified water @ 80 °C for 30 minutes

sterility

(May be steam sterilized for 3 steam-in-place (SIP) cycles of 30 minutes @ 123 °C)

feature

hydrophilic

conductivity measuring range

5-7 μmho/cm

parameter

0.03 bar max. differential pressure (0.5 psid) (Reverse)
0.21 bar max. differential pressure (3 psid) at 123 °C (Forward)
1.03 bar max. differential pressure (15 psid) at 80 °C (Forward)
2.07 bar max. differential pressure (30 psid) at 25 °C (Forward)

H

5.94 cm (2.34 in.)

diam.

16 in. , 2-cell
40.64 cm (16 in.)

filtration area

0.45 m2

impurities

<0.25 EU/mL Aqueous extraction (LAL test)

total organic carbon (TOC) residue

565-1037 ppb

filter grade

CE25
DE40

General description

after a pure water flush of 10 L/ft2 (100 L/m2) of media surface area per autoclaved (1 cycle of 30 minutes at 123 °C) 12" 16-stack device
post autoclave (1 cycle of 60 minutes at 123 °C) and pure water flush of 10 liters per ft2 (100 L/m2) of media surface area

Analysis Note

The effluent of an autoclaved (1 cycle of 30 minutes @ 123 °C) stacked device is negative after a WFI flush of 10 liters per ft2 (100 L/m2) of media surface area

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1

target_organs

Lungs

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service