Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LUXGCS3EXWB

Millipore

MPAGE®® Lux Warranty

3-year extended warranty for LUXGCS (mPAGE® Lux Curing Station), purchase at time-of-sale

Synonym(s):

Lux, Warranty

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105312
NACRES:
NA.32
Pricing and availability is not currently available.

Product Name

3-year extended warranty B for mPAGE® Lux Curing Station, 3-year extended warranty for LUXGCS (mPAGE® Lux Curing Station), purchase at time-of-sale

product line

mPAGE®

technique(s)

SDS-PAGE: suitable
electrophoresis: suitable

compatibility

mPAGE® LUXGCS

General description

3-year extended warranty for LUXGCS (mPAGE® Lux Curing Station), purchased after sale. The LUXGCS comes with a one-year warranty. Additional warranty can be purchased separately up to two years after sales. This product extends the warranty to cover a total of five years. The instrument will be replaced if broken during the warranty period.

Other Notes

This is a stand-alone product, but it can only be purchased within 2 years of purchasing mPAGE® Lux Curing Station.

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service