Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LX0331

Supelco

Lithium Chloride

Meets ACS Specifications GR ACS

Synonym(s):

Lithium Chloride, Chlorku Litu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
ClLi
CAS Number:
Molecular Weight:
42.39
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-212-3
assay:
≥99% (silver nitrate titration)
grade:
ACS reagent
form:
powder
solubility:
832 g/L

grade

ACS reagent

Quality Level

vapor pressure

1.33 hPa ( 547 °C)

assay

≥99% (silver nitrate titration)

form

powder

impurities

≤0.008 meq/g Titrable base
≤0.01% Insoluble matter

loss

≤1.0% loss on drying, 105°C

color

white to off-white

pH

6 (20 °C, 50 g/L in H2O)

bp

1360 °C/1013 hPa

mp

614 °C

solubility

832 g/L

anion traces

sulfate (SO42-): ≤0.01%

cation traces

Ba: ≤0.003%
Ca: ≤0.01%
Fe: ≤0.001%
K: ≤0.01%
Na: ≤0.20%
heavy metals: ≤0.002%

shipped in

ambient

storage temp.

room temp

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Application

Meets ACS Specifications GR ACS

Analysis Note

Assay (argentometric)99% minBarium (Ba)0.003% maxCalcium (Ca)0.01% maxColorWhite to off-whiteFormPowder or fine granulesHeavy Metals (by ICP-OES)0.002% maxInsoluble matter0.01% maxIron (Fe)0.001% maxLoss on drying (105°C)1.0% maxPotassium (K)0.01% maxSodium (Na)0.20% maxSulfate (SO4)0.01% maxTitrable base0.008meq/g max

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service