MA0HC01FS1
TPE (Thermo Plastic Elastomer)
TPE gasket (Thermo Plastic Elastomer)
cellulose membrane (and inorganic filter aid)
polypropylene housing (glass-filled)
autoclavable compatible
0.11 m2
seal (flat seal)
(A0HC media series)
1 of 4
This Item | MA0HC10FS1 | MDE3507FS1 | MA0HC23CL3 |
---|---|---|---|
fitting seal (flat seal) | fitting seal (flat seal) | fitting seal (flat seal) | fitting seal (flat seal) |
cartridge code (A0HC media series) | cartridge code (A0HC media series) | cartridge code DE35 (media series) | cartridge code (A0HC media series) |
sterilization compatibility autoclavable compatible | sterilization compatibility autoclavable compatible | sterilization compatibility autoclavable compatible | sterilization compatibility autoclavable compatible |
effective filtration area 0.11 m2 | effective filtration area 1.1 m2 | effective filtration area 0.77 m2 | effective filtration area 23 m2 |
Quality Level 400 | Quality Level 400 | Quality Level 400 | Quality Level 400 |
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
How to Find the Product Number
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).
Example:
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.
How to Find a Lot/Batch Number for COA
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').
For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').
For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.
In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.