MAB131886
Anti-DLVPR Tag Antibody, clone G196
clone G196
Sign Into View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available.
About This Item
UNSPSC Code:
12352203
Skip To
clone
G196, monoclonal
isotype
IgG1κ
Immunogen
Glutathione S-transferase (GST) bacterially expressed using the pGEX-2T vector.
Biochem/physiol Actions
Clone G196 is a mouse monoclonal antibody that detects SLVPR tag in proteins. It targets an epitope within five amino acids (DLVPR) in the C-terminal extension of GST, but does not recognize the core GST.
Preparation Note
0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Stable for 1 year at 2-8°C from date of receipt.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service